missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1H-indazole, 97%, Thermo Scientific Chemicals

Catalog Number p-7032093
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
874-05-5
C7H7N3
133.154
MFCD00182045
YDTDKKULPWTHRV-UHFFFAOYSA-N
3-amino-1h-indazole, 1h-indazol-3-ylamine, 3-aminoindazole, 3-indazolamine, 2h-indazol-3-amine, 1h-indazole, 3-amino, aminoindazole, amino-indazole, 1h-indazole-3-ylamine, indazole amine
13399
1H-indazol-3-amine
C1=CC=C2C(=C1)C(=NN2)N
This item is not returnable. View return policy
874-05-5
C7H7N3
133.154
MFCD00182045
YDTDKKULPWTHRV-UHFFFAOYSA-N
3-amino-1h-indazole, 1h-indazol-3-ylamine, 3-aminoindazole, 3-indazolamine, 2h-indazol-3-amine, 1h-indazole, 3-amino, aminoindazole, amino-indazole, 1h-indazole-3-ylamine, indazole amine
13399
1H-indazol-3-amine
C1=CC=C2C(=C1)C(=NN2)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 150°C to 152°C
Quantity 1 g
UN Number UN2811
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 3-Amino-1H-indazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'