missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Amino-1-propanol, Sigma-Aldrich™

Artikelnummer. 15893991
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15893991 250 mL
15883991 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15893991 Leverandør Sigma Aldrich Leverandørnr. 8004230250
 Begrænset vare
Forventet forsendelse: 26-11-2026
Læg i indkøbskurv
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-87-6
Grad Research
Molekylvægt (g/mol) 75.11

Specifications

Smeltepunkt 11.5°C to 12.4°C
Tæthed 0.99 g/cm3 at 20°C
Kogepunkt 187°C to 190°C
MDL nummer MFCD00008223
Mængde 250 mL
Synonym 3-Amino-1-propanol, 3-Amino-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.