Få mere at vide
3-Amino-1-propanol-d6, TRC
Artikelnummer. 30312325
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30312325 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312325 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6281332.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Amino-1-propanol-d6
CAS 59720-07-9
Molekylær formel C3 D6 H3 N O
Mængde 2.5 mg
Synonym 1-Propan-1,1,2,2,3,3-d6-ol, 3-amino- (9CI), 1-Hydroxy-3-aminopropane-1,1,2,2,3,3-d6, 3-Amino-1-propanol-1,1,2,2,3,3-d6, 3-Aminopropanol-d6, 3-Aminopropanol-D6, 3-Aminopropyl-d6 alcohol
InChI formel InChI=1S/C3H9NO/c4-2-1-3-5/h5H,1-4H2/i1D2,2D2,3D2
SMIL [2H]C([2H])(N)C([2H])([2H])C([2H])([2H])O
IUPAC navn 3-amino-1,1,2,2,3,3-hexadeuteriopropan-1-ol
Molekylvægt (g/mol) 81.1466
Formel vægt 81.1061