Learn More
3-Amino-1,2-propanediol, Sigma-Aldrich™
Product Code. 15854421
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
Product Code. Quantity Unit Size
15854421 25 mL 25mL
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15854421 Supplier Sigma Aldrich Supplier No. 8185220025
 Restricted Item
Forventet forsendelse: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 616-30-8
Density 1.18 g/cm3 at 20°C
Molecular Formula NH2CH2CH(OH)CH2OH
MDL Number MFCD00008140
Quantity 25 mL
Synonym 3-Amino-1,2-propanediol, 1-Amino-2,3-propanediol, 3-Amino-1,2-propanediol
Molecular Weight (g/mol) 91.11
Grade Research