Få mere at vide
3-Amino-1,2,4-triazole-d4, CDN
Artikelnummer. 30315728
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315728 0.1 g 0.1g
30326587 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315728 Leverandør CDN Leverandørnr. CDND82290.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Amitrole D4
CAS 787577-24-6
Molekylær formel C2 D4 N4
Mængde 0.1 g
Synonym 1 H-1,2,4-Triazol-1,5-d2-3-amine-d2 (9 CI), Amitrole-d4, Amitrole D4, 3-Amino-1,2,4-triazole-d4
InChI formel InChI=1 S/C2H4N4/c3-2-4-1-5-6-2/h1H,(H3,3,4,5,6)/i1D/hD3
SMIL [2 H]N([2 H])c1nc([2 H])n([2 H])n1
IUPAC navn N,N,1,5-tetradeuterio-1,2,4-triazol-3-amine
Molekylvægt (g/mol) 88.1046
Formel vægt 88.0687 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity