Få mere at vide
3-Acetyl-2-methylquinoxaline, TRC
Artikelnummer. 30322815
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322815 250 mg
30311692 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30322815 Leverandør Toronto Research Chemicals Leverandørnr. TRCA186875250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Acetyl-2-methylquinoxaline
CAS 22059-64-9
Molekylær formel C11H10N2O
Mængde 250 mg
Synonym 1-(3-Methyl-2-quinoxalinyl)ethanone, 2-Acetyl-3-methylquinoxaline
InChI formel InChI=1S/C11H10N2O/c1-7-11(8(2)14)13-10-6-4-3-5-9(10)12-7/h3-6H,1-2H3
SMIL CC(=O)c1nc2ccccc2nc1C
IUPAC navn 1-(3-methylquinoxalin-2-yl)ethanone
Molekylvægt (g/mol) 186.21
Formel vægt 186.0793