Få mere at vide
3-Acetyl-17-deacetyl Rocuronium Bromide, TRC
Artikelnummer. 30011369
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011369 100 mg
30008955 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011369 Leverandør Toronto Research Chemicals Leverandørnr. TRCA172180100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1190105-63-5
Molekylær formel C32 H53 N2 O4 . Br
Molekylvægt (g/mol) 609.68
Synonym Pyrrolidinium, 1-[(2β,3α,5α,16β,17β)-3-(acetyloxy)-17-hydroxy-2-(4-morpholinyl)androstan-16-yl]-1-(2-propen-1-yl)-, bromide (1:1), 1-[3α-Acetoxy-17β-hydroxy-2β-(morpholin-4-yl)-5α-androstan-16β-yl]-1-(prop-2-enyl)pyrrolidinium Bromide
IUPAC navn [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-17-hydroxy-10,13-dimethyl-2-morpholin-4-yl-16-(1-prop-2-enylpyrrolidin-1-ium-1-yl)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate;bromide
SMIL [Br-].CC(=O)O[C@H]1C[C@@H]2CC[C@@H]3[C@H](CC[C@]4(C)[C@@H](O)[C@H](C[C@@H]34)[N+]5(CC=C)CCCC5)[C@@]2(C)C[C@@H]1N6CCOCC6
Tekniske data
Kemisk navn eller materiale 3-Acetyl-17-deacetyl Rocuronium Bromide
Analyt- eller komponentnavne 3-Acetyl-17-deacetyl Rocuronium Bromide
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C32H53N2O4.BrH/c1-5-14-34(15-6-7-16-34)28-20-26-24-9-8-23-19-29(38-22(2)35)27(33-12-17-37-18-13-33)21-32(23,4)25(24)10-11-31(26,3)30(28)36;/h5,23-30,36H,1,6-21H2,2-4H3;1H/q+1;/p-1/t23-,24+,25-,26-,27-,28-,29-,30-,31-,32-;/m0./s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 100 mg
Formel vægt 608.3189
Procent renhed >95%