missing translation for 'onlineSavingsMsg'
Learn More

3-(9-Carbazolyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog Number 15482299
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
864377-33-3
C18H14BNO2
287.125
MFCD18072506
IDQUIFLAFFZYEX-UHFFFAOYSA-N
3-9h-carbazol-9-yl phenylboronic acid, 3-9h-carbazol-9-yl phenyl boronic acid, boronic acid, 3-9h-carbazol-9-yl phenyl, 3-9-carbazolyl benzeneboronic acid, 3-carbazol-9-yl phenylboronic acid, 3-9h-9-carbozale phenylboronic acid, 3-9h-9-carbazole phenylboronic acid, 3-9h-carbazol-9-yl benzeneboronic acid
56965778
(3-carbazol-9-ylphenyl)boronic acid
B(C1=CC(=CC=C1)N2C3=CC=CC=C3C4=CC=CC=C42)(O)O
This item is not returnable. View return policy
864377-33-3
C18H14BNO2
287.125
MFCD18072506
IDQUIFLAFFZYEX-UHFFFAOYSA-N
3-9h-carbazol-9-yl phenylboronic acid, 3-9h-carbazol-9-yl phenyl boronic acid, boronic acid, 3-9h-carbazol-9-yl phenyl, 3-9-carbazolyl benzeneboronic acid, 3-carbazol-9-yl phenylboronic acid, 3-9h-9-carbozale phenylboronic acid, 3-9h-9-carbazole phenylboronic acid, 3-9h-carbazol-9-yl benzeneboronic acid
56965778
(3-carbazol-9-ylphenyl)boronic acid
B(C1=CC(=CC=C1)N2C3=CC=CC=C3C4=CC=CC=C42)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Beilstein 15530380
Formula Weight 287.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-(9-Carbazolyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'