Få mere at vide
3-(9,10-Ethanoanthracen-9(10H)-yl)-N,N-dimethylpropan-1-amine Hydrochloride, Mikromol™
Beskrivelse
3-(9,10-Ethanoanthracen-9(10H)-yl)-N,N-dimethylpropan-1-amine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-(9,10-Ethanoanthracen-9(10H)-yl)-N,N-dimethylpropan-1-amine Hydrochloride |
| CAS | 23941-45-9 |
| Analyt- eller komponentnavne | 3-(9,10-Ethanoanthracen-9(10H)-yl)-N,N-dimethylpropan-1-amine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H25 N . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Maprotiline Hydrochloride Imp. E (EP), Maprotiline Imp. E (EP), 3-(9,10-Ethanoanthracen-9(10H)-yl)-N,N-dimethylpropan-1-amine Hydrochloride, Maprotiline Hydrochloride Imp. E (EP) as Hydrochloride, Maprotiline Hydrochloride Impurity E as Hydrochloride, Maprotiline Impurity E as Hydrochloride |
| InChI formel | InChI=1S/C21H25N.ClH/c1-22(2)15-7-13-21-14-12-16(17-8-3-5-10-19(17)21)18-9-4-6-11-20(18)21;/h3-6,8-11,16H,7,12-15H2,1-2H3;1H |
| SMIL | Cl.CN(C)CCCC12CCC(c3ccccc13)c4ccccc24 |
| IUPAC navn | 3-(9,10-Ethanoanthracen-9(10H)-yl)-N,N-dimethylpropan-1-amine hydrochloride |
| Vis mere |