Få mere at vide
3-(9,10-Ethanoanthracen-9(10H)-yl)-N-methylprop-2-en-1-amine Hydrochloride (Dehydromaprotiline Hydrochloride), Mikromol™
Beskrivelse
3-(9,10-Ethanoanthracen-9(10H)-yl)-N-methylprop-2-en-1-amine Hydrochloride (Dehydromaprotiline Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-(9,10-Ethanoanthracen-9(10H)-yl)-N-methylprop-2-en-1-amine Hydrochloride (Dehydromaprotiline Hydrochloride) |
| CAS | 38849-14-8 |
| Analyt- eller komponentnavne | 3-(9,10-Ethanoanthracen-9(10H)-yl)-N-methylprop-2-en-1-amine Hydrochloride (Dehydromaprotiline Hydrochloride) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H21 N . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Maprotiline Hydrochloride Imp. D (EP), Maprotiline Imp. D (EP), Maprotiline USP Related Compound D, Maprotiline USP RC D, 3-(9,10-Ethanoanthracen-9(10H)-yl)-N-methylprop-2-en-1-amine Hydrochloride, Maprotiline Hydrochloride Imp. D (EP) as Hydrochloride, Dehydromaprotiline Hydrochloride, Maprotiline Related Compound D as Hydrochloride, Maprotiline Hydrochloride Impurity D as Hydrochloride, Maprotiline Impurity D as Hydrochloride |
| InChI formel | InChI=1S/C20H21N.ClH/c1-21-14-6-12-20-13-11-15(16-7-2-4-9-18(16)20)17-8-3-5-10-19(17)20;/h2-10,12,15,21H,11,13-14H2,1H3;1H/b12-6+; |
| SMIL | Cl.CNC\C=C\C12CCC(c3ccccc13)c4ccccc24 |
| IUPAC navn | 3-(9,10-Ethanoanthracen-9(10H)-yl)-N-methylprop-2-en-1-amine hydrochloride |
| Vis mere |