Få mere at vide
3-(7-Cyano-5-(2-nitropropyl)indolin-1-yl)propyl Benzoate, TRC
Artikelnummer. 30362216
Skift visning
Mængde:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362216 5 g
30364125 100 g
30353585 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30362216 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9820355G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 350797-56-7
Molekylær formel C22H23N3O4
Molekylvægt (g/mol) 393.44
Synonym 1-[3-(Benzoyloxy)propyl]-2,3-dihydro-5-(2-nitropropyl)-1H-Indole-7-carbonitrile
IUPAC navn 3-[7-cyano-5-(2-nitropropyl)-2,3-dihydroindol-1-yl]propyl benzoate
SMIL CC(Cc1cc2CCN(CCCOC(=O)c3ccccc3)c2c(c1)C#N)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 3-(7-Cyano-5-(2-nitropropyl)indolin-1-yl)propyl Benzoate
Mængde 5 g
InChI formel InChI=1S/C22H23N3O4/c1-16(25(27)28)12-17-13-19-8-10-24(21(19)20(14-17)15-23)9-5-11-29-22(26)18-6-3-2-4-7-18/h2-4,6-7,13-14,16H,5,8-12H2,1H3
Formel vægt 393.1689