missing translation for 'onlineSavingsMsg'
Learn More

3(6R,7R)-Hydroxy-8-oxo-7-[(2-phenylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid Diphenylmethyl Ester (>90%), TRC

Product Code. 30405607
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30405607 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405607 Supplier Toronto Research Chemicals Supplier No. TRCH94948550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3(6R,7R)-Hydroxy-8-oxo-7-[(2-phenylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid Diphenylmethyl Ester
CAS 54639-48-4
Molekylær formel C28H24N2O5S
Mængde 50 mg
Synonym 3-Hydroxy-8-oxo-7-[(phenylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid Diphenylmethyl Ester, (6R-trans)-3-Hydroxy-8-oxo-7-[(phenylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Diphenylmethyl Ester, 7-Phenylacetamido-3-hydroxy-3-cephem-4-carboxylic Acid Diphenylmethyl Ester
InChI formel InChI=1S/C28H24N2O5S/c31-21-17-36-27-23(29-22(32)16-18-10-4-1-5-11-18)26(33)30(27)24(21)28(34)35-25(19-12-6-2-7-13-19)20-14-8-3-9-15-20/h1-15,23,25,27,31H,16-17H2,(H,29,32)/t23-,27-/m1/s1
SMIL OC1=C(C(OC(C2=CC=CC=C2)C3=CC=CC=C3)=O)N4[C@]([C@H](NC(CC5=CC=CC=C5)=O)C4=O)([H])SC1
IUPAC navn benzhydryl (6R,7R)-3-hydroxy-8-oxo-7-[(2-phenylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molekylvægt (g/mol) 500.57
Formel vægt 500.1406
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.