missing translation for 'onlineSavingsMsg'
Learn More

3-(6-Methyl-3-pyridinyl)propanoic Acid, TRC

Product Code. 30480402
Change view
Click to view available options
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30480402 10 mg
30482104 100 mg
30470541 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480402 Supplier Toronto Research Chemicals Supplier No. TRCM34000010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118420-23-8
Molekylær formel C9H11NO2
Molekylvægt (g/mol) 165.19
IUPAC navn 3-(6-methylpyridin-3-yl)propanoic acid
SMIL Cc1ccc(CCC(=O)O)cn1

Specifications

Kemisk navn eller materiale 3-(6-Methyl-3-pyridinyl)propanoic Acid
Mængde 10 mg
InChI formel InChI=1S/C9H11NO2/c1-7-2-3-8(6-10-7)4-5-9(11)12/h2-3,6H,4-5H2,1H3,(H,11,12)
Formel vægt 165.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.