Få mere at vide
3-(6-Methoxy-2-naphthalenyl)-5-methylcyclohexanone (Impurity), TRC
Artikelnummer. 30011955
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011955 250 mg
30004068 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011955 Leverandør Toronto Research Chemicals Leverandørnr. TRCM264770250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 343272-52-6
Molekylær formel C18 H20 O2
Molekylvægt (g/mol) 268.35
Synonym Cyclohexanone, 3-(6-methoxy-2-naphthalenyl)-5-methyl-, 3-(6-Methoxynaphthalen-2-yl)-5-methylcyclohexanone, Nabumetone Imp. A (EP)
IUPAC navn 3-(6-methoxynaphthalen-2-yl)-5-methylcyclohexan-1-one
SMIL COc1ccc2cc(ccc2c1)C3CC(C)CC(=O)C3
Tekniske data
Kemisk navn eller materiale 3-(6-Methoxynaphthalen-2-yl)-5-methylcyclohexanone
Mængde 250 mg
InChI formel InChI=1S/C18H20O2/c1-12-7-16(10-17(19)8-12)14-3-4-15-11-18(20-2)6-5-13(15)9-14/h3-6,9,11-12,16H,7-8,10H2,1-2H3
Formel vægt 268.1463