Få mere at vide
3,6-Dioxo-19-nor-17alpha-pregn-4-en-20-yn-17-yl Acetate (6-Ketonorethisterone Acetate), Mikromol™
Artikelnummer. 30052121
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052121 25 mg
30051814 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052121 Leverandør Mikromol Leverandørnr. MM0219.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3,6-Dioxo-19-nor-17alpha-pregn-4-en-20-yn-17-yl Acetate (6-Ketonorethisterone Acetate)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,6-Dioxo-19-nor-17alpha-pregn-4-en-20-yn-17-yl Acetate (6-Ketonorethisterone Acetate)
CAS 438244-27-0
Analyt- eller komponentnavne 3,6-Dioxo-19-nor-17alpha-pregn-4-en-20-yn-17-yl Acetate (6-Ketonorethisterone Acetate)
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C22H26O4
Certificeringer/compliance ISO 17025
Synonym Norethisterone Acetate Imp. G (EP), 3,6-Dioxo-19-nor-17alpha-pregn-4-en-20-yn-17-yl Acetate, 6-Ketonorethisterone Acetate, Norethisterone Acetate Impurity G
SMIL CC(=O)O[C@]1(CC[C@H]2[C@@H]3CC(=O)C4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C
Molekylvægt (g/mol) 354.44