Få mere at vide
3,6-Dimethyl-3-heptanol, TRC
Artikelnummer. 30376654
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376654 250 mg
30375233 100 mg
30376551 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30376654 Leverandør Toronto Research Chemicals Leverandørnr. TRCD473110250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1573-28-0
Molekylær formel C9 H20 O
Molekylvægt (g/mol) 144.25
Synonym 3,6-Dimethylheptan-3-ol
IUPAC navn 3,6-dimethylheptan-3-ol
SMIL CCC(C)(O)CCC(C)C
Tekniske data
Kemisk navn eller materiale 3,6-Dimethyl-3-heptanol
Mængde 250 mg
InChI formel InChI=1S/C9H20O/c1-5-9(4,10)7-6-8(2)3/h8,10H,5-7H2,1-4H3
Formel vægt 144.1514