Learn More
3',6'-Dihydroxy-6-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 18861-78-4 |
| Molecular Formula | C21H11NO5S |
| MDL Number | MFCD00041838 |
| Quantity | 100 mg |
| SMILES | O=C1OC2(C3=C(OC4=C2C=CC(O)=C4)C=C(O)C=C3)C5=C1C=CC(N=C=S)=C5 |
| Molecular Weight (g/mol) | 389.38 |
| Percent Purity | 95% |
| Chemical Name or Material | 3',6'-Dihydroxy-6-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one |