missing translation for 'onlineSavingsMsg'
Få mere at vide

3',6'-Dihydroxy-5-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 95%, BLD Pharm™

Artikelnummer. 30967942
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 g
5 g
Pakningsstørrelse:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
30967942 1 g 1g
30964662 5 g 5g
30964034 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30967942 Leverandør BLD Pharm Leverandørnr. BD12901g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale 3',6'-Dihydroxy-5-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one
CAS 3326-32-7
Molekylær formel C21H11NO5S
MDL nummer MFCD00005063
Mængde 1 g
SMIL O=C1OC2(C3=C(OC4=C2C=CC(O)=C4)C=C(O)C=C3)C5=C1C=C(N=C=S)C=C5
Molekylvægt (g/mol) 389.38
Procent renhed 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.