missing translation for 'onlineSavingsMsg'
Learn More

3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 98%, BLD Pharm™

Product Code. 30968022
Encompass_Preferred
Change view
Click to view available options
Mængde:
100 mg
250 mg
1 g
5 g
10 g
25 g
Unit Size:
100mg
10g
1g
250mg
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde Unit Size
30968022 10 g 10g
30962093 250 mg 250mg
30949096 25 g 25g
30956878 100 mg 100mg
30952002 1 g 1g
30951863 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30968022 Supplier BLD Pharm Supplier No. BD10860210g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid
CAS 3301-79-9
Molekylær formel C21H12O7
MDL nummer MFCD00036873
Mængde 10 g
SMIL O=C1OC2(C3=C(OC4=C2C=CC(O)=C4)C=C(O)C=C3)C5=C1C=CC(C(O)=O)=C5
Molekylvægt (g/mol) 376.32
Procent renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.