missing translation for 'onlineSavingsMsg'
Learn More

3,6-Di-2-pyridyl-1,2,4,5-tetrazine, 96%, Thermo Scientific Chemicals

Catalog Number p-7035757
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1671-87-0
C12H8N6
236.24
MFCD00121717
JFBIRMIEJBPDTQ-UHFFFAOYSA-N
3,6-di pyridin-2-yl-1,2,4,5-tetrazine, 3,6-di 2-pyridyl-1,2,4,5-tetrazine, 3,6-di-2-pyridyl-1,2,4,5-tetrazine, 3,6-di-2-pyridinyl-1,2,4,5-tetrazine, 3,6-bis 2-pyridyl-1,2,4,5-tetrazine, bis pyridin-2-yl-1,2,4,5-tetrazine, 3,6-bis 2-pyridinyl-1,2,4,5-tetrazine, bptz, acmc-209dvt, 3,6-di-2-pyridyl-s-tetrazine
315109
3,6-dipyridin-2-yl-1,2,4,5-tetrazine
C1=CC=C(N=C1)C1=NN=C(N=N1)C1=CC=CC=N1
This item is not returnable. View return policy
1671-87-0
C12H8N6
236.24
MFCD00121717
JFBIRMIEJBPDTQ-UHFFFAOYSA-N
3,6-di pyridin-2-yl-1,2,4,5-tetrazine, 3,6-di 2-pyridyl-1,2,4,5-tetrazine, 3,6-di-2-pyridyl-1,2,4,5-tetrazine, 3,6-di-2-pyridinyl-1,2,4,5-tetrazine, 3,6-bis 2-pyridyl-1,2,4,5-tetrazine, bis pyridin-2-yl-1,2,4,5-tetrazine, 3,6-bis 2-pyridinyl-1,2,4,5-tetrazine, bptz, acmc-209dvt, 3,6-di-2-pyridyl-s-tetrazine
315109
3,6-dipyridin-2-yl-1,2,4,5-tetrazine
C1=CC=C(N=C1)C1=NN=C(N=N1)C1=CC=CC=N1

3,6-Di-2-pyridyl-1,2,4,5-tetrazine may be used in the preparation of mononuclear, cyclic tetranuclear, and 1-D helical-chain Cu(II) complexes, via metal-assisted hydrolysis. It also may be used in the synthesis of novel cyclic tetranuclear ZnII complex and synthesis of substituted 3,6-di(2-pyridyl)pyridazines, via inverse electron demand Diels Alder reaction with alkenes or alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,6-Di-2-pyridyl-1,2,4,5-tetrazine may be used in the preparation of mononuclear, cyclic tetranuclear, and 1-D helical-chain Cu(II) complexes, via metal-assisted hydrolysis. It also may be used in the synthesis of novel cyclic tetranuclear ZnII complex and synthesis of substituted 3,6-di(2-pyridyl)pyridazines, via inverse electron demand Diels Alder reaction with alkenes or alkynes.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 225°C (decomposition)
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 236.24
Percent Purity 96%
Chemical Name or Material 3,6-Di-2-pyridyl-1,2,4,5-tetrazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'