Få mere at vide
3,6,9,12,15-Pentaoxaheptadecane-1,17-diyl Bis-methanethiosulfonate (>90%), TRC
Artikelnummer. 30427792
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427792 25 mg
30422547 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30427792 Leverandør Toronto Research Chemicals Leverandørnr. TRCP27375025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 384342-61-4
Molekylær formel C14 H30 O9 S4
Molekylvægt (g/mol) 470.64
Synonym 3,6,9,12,15-Pentaoxaheptadecane-1,17-Diyl Bis-Methanethiosulfonate
IUPAC navn 1-[2-(2-methylsulfonylsulfanylethoxy)ethoxy]-2-[2-[2-(2-methylsulfonylsulfanylethoxy)ethoxy]ethoxy]ethane
SMIL CS(=O)(=O)SCCOCCOCCOCCOCCOCCSS(=O)(=O)C
Tekniske data
Kemisk navn eller materiale 3,6,9,12,15-Pentaoxaheptadecane-1,17-diyl Bis-methanethiosulfonate
Mængde 25 mg
InChI formel InChI=1S/C14H30O9S4/c1-26(15,16)24-13-11-22-9-7-20-5-3-19-4-6-21-8-10-23-12-14-25-27(2,17)18/h3-14H2,1-2H3
Formel vægt 470.0773