Få mere at vide
3-(5H-Dibenzo[a,d][7]annulen-5-ylidene)-N-methylpropan-1-amine Hydrochloride (Norcyclobenzaprine Hydrochloride), Mikromol™
Beskrivelse
3-(5H-Dibenzo[a,d][7]annulen-5-ylidene)-N-methylpropan-1-amine Hydrochloride (Norcyclobenzaprine Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Desmethylcyclobenzaprine HCl |
| CAS | 438-59-5 |
| Analyt- eller komponentnavne | N-Desmethylcyclobenzaprine HCl |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H19 N . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Nortriptyline Hydrochloride Imp. B (EP), Nortriptyline Imp. B (EP), Cyclobenzaprine USP Related Compound B, Cyclobenzaprine USP RC B, 3-(5H-Dibenzo-[a,d][7]annulen-5-ylidene)-N-methylpropan-1-amine Hydrochloride, Nortriptyline Hydrochloride Imp. B (EP) as Hydrochloride, Norcyclobenzaprine Hydrochloride, Cyclobenzaprine Related Compound B as Hydrochloride, Nortriptyline Hydrochloride Impurity B as Hydrochloride, Nortriptyline Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C19H19N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-13,20H,6,14H2,1H3;1H |
| SMIL | Cl.CNCCC=C1c2ccccc2C=Cc3ccccc13 |
| IUPAC navn | 3-(dibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)-N-methylpropan-1-amine;hydrochloride |
| Vis mere |