missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzenesulfonyl chloride, Tech., Thermo Scientific™

Catalog Number p-3110380
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
10185-62-3
C9H7ClN2O3S
258.68
MFCD06797486
YPMVPGZXUILSPT-UHFFFAOYSA-N
3-5-methyl-1,2,4-oxadiazol-3-yl benzenesulfonyl chloride, benzenesulfonylchloride,3-5-methyl-1,2,4-oxadiazol-3-yl, 3-5-methyl-1,2,4-oxadiazol-3-yl benzene-1-sulfonyl chloride, chloro 3-5-methyl 1,2,4-oxadiazol-3-yl phenyl sulfone, 3-5-methyl-1,2,4 oxadiazol-3-yl-benzenesulfonylchloride, 3-5-methyl-1,2,4-oxadiazol-3-yl phenylsulfonyl chloride, benzenesulfonyl chloride, 3-5-methyl-1,2,4-oxadiazol-3-yl, benzenesulfonylchloride, 3-5-methyl-1,2,4-oxadiazol-3-yl
18525745
CC1=NC(=NO1)C1=CC(=CC=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
10185-62-3
C9H7ClN2O3S
258.68
MFCD06797486
YPMVPGZXUILSPT-UHFFFAOYSA-N
3-5-methyl-1,2,4-oxadiazol-3-yl benzenesulfonyl chloride, benzenesulfonylchloride,3-5-methyl-1,2,4-oxadiazol-3-yl, 3-5-methyl-1,2,4-oxadiazol-3-yl benzene-1-sulfonyl chloride, chloro 3-5-methyl 1,2,4-oxadiazol-3-yl phenyl sulfone, 3-5-methyl-1,2,4 oxadiazol-3-yl-benzenesulfonylchloride, 3-5-methyl-1,2,4-oxadiazol-3-yl phenylsulfonyl chloride, benzenesulfonyl chloride, 3-5-methyl-1,2,4-oxadiazol-3-yl, benzenesulfonylchloride, 3-5-methyl-1,2,4-oxadiazol-3-yl
18525745
CC1=NC(=NO1)C1=CC(=CC=C1)S(Cl)(=O)=O
CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 3-(5-methyl-1,2,4-oxadiazol-3-yl)benzenesulfonyl chloride
Formula Weight 258.68
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-(5-Methyl-1, 2, 4-oxadiazol-3-yl)benzenesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'