missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzotrifluoride, 98%, Thermo Scientific Chemicals

Catalog Number p-7027470
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
401-99-0
C7H3F3N2O4
236.106
MFCD00007233
QZADIXWDDVQVKM-UHFFFAOYSA-N
3,5-dinitrobenzotrifluoride, 1,3-dinitro-5-trifluoromethyl benzene, benzene, 1,3-dinitro-5-trifluoromethyl, 3,5-dinitrotrifluorotoluene, alpha,alpha,alpha-trifluoro-3,5-dinitrotoluene, pubchem10396, 3, 5-dinitrotrifluorotoluene, 3,5-dinitro-benzotrifluoride, 3,5-?dinitrobenzotrifluoride
123060
1,3-dinitro-5-(trifluoromethyl)benzene
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(F)(F)F
This item is not returnable. View return policy
401-99-0
C7H3F3N2O4
236.106
MFCD00007233
QZADIXWDDVQVKM-UHFFFAOYSA-N
3,5-dinitrobenzotrifluoride, 1,3-dinitro-5-trifluoromethyl benzene, benzene, 1,3-dinitro-5-trifluoromethyl, 3,5-dinitrotrifluorotoluene, alpha,alpha,alpha-trifluoro-3,5-dinitrotoluene, pubchem10396, 3, 5-dinitrotrifluorotoluene, 3,5-dinitro-benzotrifluoride, 3,5-?dinitrobenzotrifluoride
123060
1,3-dinitro-5-(trifluoromethyl)benzene
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 50°C to 52°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2058079
Formula Weight 236.11
Percent Purity 98%
Chemical Name or Material 3,5-Dinitrobenzotrifluoride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'