Learn More
3,5-Dimethylisoxazole-4-carbonyl chloride, 98%, Thermo Scientific™
Catalog Number p-7033562
Click to view available options
Quantity:
1 g
5 g
31301-45-8
C6H6ClNO2
159.569
MFCD00052555
MPYGFFPGJMGVSW-UHFFFAOYSA-N
3,5-dimethylisoxazole-4-carbonyl chloride, 4-isoxazolecarbonyl chloride, 3,5-dimethyl, 3,5-dimethylisoxazole-4-carbonylchloride, dimethyl-1,2-oxazole-4-carbonyl chloride, 3,5-dimethyl-4-isoxazolecarbonyl chloride, pubchem8669, acmc-20a3wc, 3,5-dimethylisoxasole-4-carbonyl chloride, 3,5-dimethyisoxazole-4-carbonyl chloride, 3,5 dimethylisoxazole-4-carbonyl chloride
2736265
3,5-dimethyl-1,2-oxazole-4-carbonyl chloride
CC1=C(C(=NO1)C)C(=O)Cl
31301-45-8
C6H6ClNO2
159.569
MFCD00052555
MPYGFFPGJMGVSW-UHFFFAOYSA-N
3,5-dimethylisoxazole-4-carbonyl chloride, 4-isoxazolecarbonyl chloride, 3,5-dimethyl, 3,5-dimethylisoxazole-4-carbonylchloride, dimethyl-1,2-oxazole-4-carbonyl chloride, 3,5-dimethyl-4-isoxazolecarbonyl chloride, pubchem8669, acmc-20a3wc, 3,5-dimethylisoxasole-4-carbonyl chloride, 3,5-dimethyisoxazole-4-carbonyl chloride, 3,5 dimethylisoxazole-4-carbonyl chloride
2736265
3,5-dimethyl-1,2-oxazole-4-carbonyl chloride
CC1=C(C(=NO1)C)C(=O)Cl
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3,5-Dimethylisoxazole-4-carbonyl chloride is used as pharmaceutical intermediates.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas. Incompatible with bases, oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 159.57
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylisoxazole-4-carbonyl chloride