Få mere at vide
3,5-Dimethylbenzoic-d9 Acid, TRC
Artikelnummer. 30375129
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30375129 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375129 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47142225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,5-Dimethylbenzoic-d9 Acid
CAS 1335014-65-7
Molekylær formel C9 D9 H O2
Mængde 25 mg
Synonym 3,5-Dimethylbenzoic-d9 acid, 3,5-Dimethylbenzoic acid-d9, Mesitylenic acid-d9, 3,5-Di(methyl-d3)benzoic-2,4,6-d3 acid, 3,5-Dimethylbenzoic acid D9
InChI formel InChI=1S/C9H10O2/c1-6-3-7(2)5-8(4-6)9(10)11/h3-5H,1-2H3,(H,10,11)/i1D3,2D3,3D,4D,5D
SMIL [2H]c1c(C(=O)O)c([2H])c(c([2H])c1C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2,4,6-trideuterio-3,5-bis(trideuteriomethyl)benzoic acid
Molekylvægt (g/mol) 159.23
Formel vægt 159.125