Få mere at vide
3,5-Dimethylbenzoic-d9 Acid, CDN
Artikelnummer. 30322307
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322307 1 g
30310029 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322307 Leverandør CDN Leverandørnr. CDND74361G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,5-Dimethylbenzoic-d9 Acid
CAS 1335014-65-7
Molekylær formel C9 D9 H O2
Mængde 1 g
Synonym 3,5-Dimethylbenzoic-d9 acid, 3,5-Dimethylbenzoic acid-d9, Mesitylenic acid-d9, 3,5-Di(methyl-d3)benzoic-2,4,6-d3 acid, 3,5-Dimethylbenzoic acid D9
InChI formel InChI=1 S/C9H10O2/c1-6-3-7(2)5-8(4-6)9(10)11/h3-5 H,1-2H3,(H,10,11)/i1D3,2D3,3 D,4 D,5 D
SMIL [2 H]c1c(C(=O)O)c([2 H])c(c([2 H])c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,4,6-trideuterio-3,5-bis(trideuteriomethyl)benzoic acid
Molekylvægt (g/mol) 159.23
Formel vægt 159.125 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity