Få mere at vide
3,5-Dimethyl-4-(methylthio)phenol, TRC
Artikelnummer. 30135414
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135414 5 g
30010399 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135414 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4748105G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7379-51-3
Molekylær formel C9 H12 O S
Molekylvægt (g/mol) 168.26
Synonym 3,5-Dimethyl-4-(methylthio)phenol, 3,5-Dimethyl-4-methylsulfanylphenol, 4-Methylsulfanyl-3,5-dimethylphenol
IUPAC navn 3,5-dimethyl-4-methylsulfanylphenol
SMIL CSc1c(C)cc(O)cc1C
Tekniske data
Kemisk navn eller materiale 3,5-Dimethyl-4-(methylthio)phenol
Mængde 5 g
InChI formel InChI=1S/C9H12OS/c1-6-4-8(10)5-7(2)9(6)11-3/h4-5,10H,1-3H3
Formel vægt 168.0609