Få mere at vide
3,5-Dihydroxynapthalene-2-carboxylic Acid, TRC
Artikelnummer. 30134414
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134414 5 g
30133898 10 g
30012531 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134414 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4535005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 89-35-0
Molekylær formel C11H8O4
Molekylvægt (g/mol) 204.18
Synonym UBP551;
SMIL OC(=O)c1cc2cccc(O)c2cc1O
Tekniske data
Kemisk navn eller materiale 3,5-Dihydroxynapthalene-2-carboxylic Acid
Mængde 5 g
InChI formel InChI=1S/C11H8O4/c12-9-3-1-2-6-4-8(11(14)15)10(13)5-7(6)9/h1-5,12-13H,(H,14,15)
Formel vægt 204.0423