Learn More
3,5-Dihydroxybenzoic Acid, Sigma-Aldrich™
Product Code. 15898380
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15898380 250 g 250g
15888380 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15898380 Supplier Sigma Aldrich Supplier No. 8007370250
 Restricted Item
Forventet forsendelse: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99-10-5
Grade Research
Molecular Weight (g/mol) 154.12
Specifications
Melting Point 234°C to 238°C
MDL Number MFCD00002512
Quantity 250 g
Synonym 3,5-Dihydroxybenzoic acid, 5-Carboxyresorcinol, 3,5-Dihydroxybenzoic acid