Få mere at vide
3,5-Difluorobenzoic-d3 Acid, CDN
Artikelnummer. 30319285
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319285 0.1 g 0.1g
30324732 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319285 Leverandør CDN Leverandørnr. CDND64860.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,5-Difluorobenzoic Acid-D3
CAS 934397-71-4
Molekylær formel C7 D3 H F2 O2
Mængde 0.1 g
Synonym Benzoic-2,4,6-d3 acid, 3,5-difluoro- (ACI), 3,5-Difluorobenzoic-2,4,6-d3 acid (ACI), 3,5-Difluorobenzoic-d3 acid, 3,5-Difluorobenzoic acid-d3, 3,5-Difluorobenzoic Acid-D3
InChI formel InChI=1 S/C7H4F2O2/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3 H,(H,10,11)/i1D,2 D,3 D
SMIL [2 H]c1c(F)c([2 H])c(C(=O)O)c([2 H])c1F
IUPAC navn 2,4,6-trideuterio-3,5-difluorobenzoic acid
Molekylvægt (g/mol) 161.1208
Formel vægt 161.0368 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity