missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-[2-fluoro-5-(trifluoromethyl)phenyl]benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62723.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15407786

  • 9607.87 DKK / 5g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Difluoro-N-[2-fluoro-5-(trifluoromethyl)phenyl]benzenesulfonamide
355.26
MFCD27665226

Specifications

3,5-Difluoro-N-[2-fluoro-5-(trifluoromethyl)phenyl]benzenesulfonamide
Solid
355.26
5g
97%
MFCD27665226
Videos
SDS