Learn More
Alfa Aesar™ 3,5-Difluoro-N-(2,4-dimethylphenyl)benzamide, 97%
Brand:  Alfa Aesar™ H61436.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15407988
  • 4807.66 DKK / 1g
Estimated Shipment date
from Supplier 07-05-2018
Add to basket
Specifications
Chemical Identifiers
3,5-Difluoro-N-(2,4-dimethylphenyl)benzamide
261.27
MFCD05474349
Specifications
3,5-Difluoro-N-(2,4-dimethylphenyl)benzamide
Solid
261.27
1g
97%
MFCD05474349