missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-methoxybenzoic acid, 97%, Thermo Scientific™

Catalog Number 15423407
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
319-60-8
C8H6F2O3
188.13
MFCD01090994
YOWQTTYAQQPSHM-UHFFFAOYSA-N
2736987
3,5-difluoro-4-methoxybenzoic acid
COC1=C(C=C(C=C1F)C(=O)O)F
This item is not returnable. View return policy
319-60-8
C8H6F2O3
188.13
MFCD01090994
YOWQTTYAQQPSHM-UHFFFAOYSA-N
2736987
3,5-difluoro-4-methoxybenzoic acid
COC1=C(C=C(C=C1F)C(=O)O)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 164°C to 166°C
Quantity 250 mg
Synonym 3,5-difluoro-4-methoxybenzoicacid, 3,5-difluoro-4-methoxy benzoic acid, 4-carboxy-2,6-difluoroanisole, benzoic acid, 3,5-difluoro-4-methoxy, 3,5-difluoro-4-methoxy-benzoic acid, acmc-20aic4, 3,5difluoro-4-methoxy benzoic acid, 3,5 difluoro-4-methoxy benzoic acid, benzoic acid,3,5-difluoro-4-methoxy, 3,5-bis fluoranyl-4-methoxy-benzoic acid
Formula Weight 188.13
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-4-methoxybenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'