Learn More
Alfa Aesar™ 3,5-Difluoro-2-methylbenzonitrile, 97%
Brand: Alfa Aesar™ H34195.03
Additional Details : CAS Number : 1003708-74-4 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
1003708-74-4 | |
153.132 | |
JHMDKULXRYFRBP-UHFFFAOYSA-N | |
44887034 | |
CC1=C(C=C(C=C1C#N)F)F |
C8H5F2N | |
MFCD09996934 | |
3,5-difluoro-2-methyl benzonitrile, pubchem8342 | |
3,5-difluoro-2-methylbenzonitrile |
Specifications
3,5-Difluoro-2-methylbenzonitrile | |
55°C to 57°C | |
1003708-74-4 | |
MFCD09996934 | |
3,5-difluoro-2-methyl benzonitrile, pubchem8342 | |
CC1=C(C=C(C=C1C#N)F)F | |
153.132 | |
153.13 | |
97% |
White | |
1g | |
C8H5F2N | |
UN3439 | |
JHMDKULXRYFRBP-UHFFFAOYSA-N | |
3,5-difluoro-2-methylbenzonitrile | |
44887034 | |
Crystalline Powder |