missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-2-methylbenzoic acid, 97%, Thermo Scientific™

Catalog Number 15444217
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1003710-06-2
C8H6F2O2
172.13
MFCD09832334
PTGLKDIYYKDEQX-UHFFFAOYSA-N
3,5-difluoro-2-methylbenzoicacid, 3,5-difluoro-2-methyl-benzoic acid, pubchem8242, 3,5-difluoro-2-methylbenzoic acid, jrd, 3,5-difluoro-2-methylbethylbenzoic acid
20111761
3,5-difluoro-2-methylbenzoic acid
CC1=C(F)C=C(F)C=C1C(O)=O
This item is not returnable. View return policy
1003710-06-2
C8H6F2O2
172.13
MFCD09832334
PTGLKDIYYKDEQX-UHFFFAOYSA-N
3,5-difluoro-2-methylbenzoicacid, 3,5-difluoro-2-methyl-benzoic acid, pubchem8242, 3,5-difluoro-2-methylbenzoic acid, jrd, 3,5-difluoro-2-methylbethylbenzoic acid
20111761
3,5-difluoro-2-methylbenzoic acid
CC1=C(F)C=C(F)C=C1C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The mineralization of acidic aqueous solutions with 230 and 115 ppm 3,6-dichloro-2-methoxybenzoic acid (dicamba) in 0.05 M Na2SO4 of pH 3.0 has been studied by electro-Fenton and photoelectro-Fenton using a Pt anode and an O2-diffusion cathode, where oxidizing hydroxyl radicals are produced from Fenton′s reaction between added Fe2+ and H2O2 generated by the cathode. The fast mineralization of such complexes by UV light explains the highest oxidative ability of photoelectro-Fenton.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 133°C to 136°C
Quantity 250 mg
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-2-methylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'