missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Difluoro-2-methylbenzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
The mineralization of acidic aqueous solutions with 230 and 115 ppm 3,6-dichloro-2-methoxybenzoic acid (dicamba) in 0.05 M Na2SO4 of pH 3.0 has been studied by electro-Fenton and photoelectro-Fenton using a Pt anode and an O2-diffusion cathode, where oxidizing hydroxyl radicals are produced from Fenton′s reaction between added Fe2+ and H2O2 generated by the cathode. The fast mineralization of such complexes by UV light explains the highest oxidative ability of photoelectro-Fenton.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
The mineralization of acidic aqueous solutions with 230 and 115 ppm 3,6-dichloro-2-methoxybenzoic acid (dicamba) in 0.05 M Na2SO4 of pH 3.0 has been studied by electro-Fenton and photoelectro-Fenton using a Pt anode and an O2-diffusion cathode, where oxidizing hydroxyl radicals are produced from Fenton′s reaction between added Fe2+ and H2O2 generated by the cathode. The fast mineralization of such complexes by UV light explains the highest oxidative ability of photoelectro-Fenton.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 133°C to 136°C |
Quantity | 1 g |
Formula Weight | 172.13 |
Percent Purity | 97% |
Chemical Name or Material | 3,5-Difluoro-2-methylbenzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'