Få mere at vide
3,5-Dichlorobenzoic-d3 Acid, TRC
Artikelnummer. 30375171
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30375171 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375171 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4365645MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,5-Dichlorobenzoic-d3 Acid
CAS 1219803-99-2
Molekylær formel C7 D3 H Cl2 O2
Mængde 5 mg
Synonym 3,5-Dichlorobenzoic-d3 Acid, 3,5-Dichlorobenzoic Acid-d3, 3,5-Dichlorobenzoic-2,4,6-d3 Acid, 3,5-Dichlorobenzoic Acid-D3
InChI formel InChI=1S/C7H4Cl2O2/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3H,(H,10,11)/i1D,2D,3D
SMIL [2H]c1c(Cl)c([2H])c(C(=O)O)c([2H])c1Cl
IUPAC navn 3,5-dichloro-2,4,6-trideuteriobenzoic acid
Molekylvægt (g/mol) 194.03
Formel vægt 192.978