Få mere at vide
3,5-Dichloro-4-formylbenzoic Acid, TRC
Artikelnummer. 30471700
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471700 50 mg
30470250 500 mg
30471508 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30471700 Leverandør Toronto Research Chemicals Leverandørnr. TRCL47498850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 153203-80-6
Molekylær formel C8H4Cl2O3
Molekylvægt (g/mol) 219.02
IUPAC navn 3,5-dichloro-4-formylbenzoic acid
SMIL OC(=O)c1cc(Cl)c(C=O)c(Cl)c1
Tekniske data
Kemisk navn eller materiale 3,5-Dichloro-4-formylbenzoic Acid
Mængde 50 mg
InChI formel InChI=1S/C8H4Cl2O3/c9-6-1-4(8(12)13)2-7(10)5(6)3-11/h1-3H,(H,12,13)
Formel vægt 217.9537