missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
3,5-dibrom-4-hydroxybenzaldehydoxim, 97 %, Thermo Scientific™
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3, 5-Dibromo-4-hydroxybenzaldehyde oxime |
| CAS | 25952-74-3 |
| CAS min % | 97.0 |
| Analyseprocentområde | 97% |
| Emballage | Ravfarvet glasflaske |
| Molekylær formel | C7H5Br2NO2 |
| MDL nummer | MFCD02180889 |
| Mængde | 1 g |
| Synonym | 3,5-dibromo-4-hydroxybenzaldehyde oxime, 2,6-dibromo-4-hydroxyamino methylidene cyclohexa-2,5-dien-1-one, 2,6-dibromo-4-hydroxyimino methyl phenol, benzaldehyde,3,5-dibromo-4-hydroxy-, oxime, 2,6-dibromo-4-1e-hydroxyimino methyl phenol, 3,5-dibromo-4-hydroxybenzaldehydeoxime, hcndpekpqnqlbbbtqscaadpbip, hcndpekpqnqlbbbtqscaadpbi`, 2,6-dibromo-4-hydroxyamino methylidene-1-cyclohexa-2,5-dienone, 2,6-bis bromanyl-4-oxidanylamino methylidene cyclohexa-2,5-dien-1-one |
| InChI nøgle | NWAZSJXQHSKZJO-UHFFFAOYSA-N |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?