missing translation for 'onlineSavingsMsg'
Learn More
3,5-Diamino-1,2,4-triazole, 98% Product Code.: 11419817

3,5-Diamino-1,2,4-triazole, 98%

Product Code. 11419817
100 g, 100g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price

Product Code. 11419817

Brand: Thermo Scientific Alfa Aesar B22775.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price

CAS: 1455-77-2 | C2H5N5 | 99.097 g/mol

3,5-Diamino-1,2,4-triazole is used as an inhibitor of DNA synthesis. It also serves as an antitumor agents in the treatment of epigenetically-based diseases. It acts as a corrosion inhibitor for copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Diamino-1,2,4-triazole is used as an inhibitor of DNA synthesis. It also serves as an antitumor agents in the treatment of epigenetically-based diseases. It acts as a corrosion inhibitor for copper.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1455-77-2
Molecular Formula C2H5N5
Molecular Weight (g/mol) 99.097
MDL Number MFCD00005233
InChI Key PKWIYNIDEDLDCJ-UHFFFAOYSA-N
Synonym guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamineShow MoreShow Less
PubChem CID 15078
ChEBI CHEBI:75425
IUPAC Name 1H-1,2,4-triazole-3,5-diamine
SMILES C1(=NC(=NN1)N)N

Specifications

Melting Point ∼205°C (decomposition)
Quantity 100 g
Beilstein 112467
Solubility Information Soluble in water.
Formula Weight 99.1
Percent Purity ≥98%
Chemical Name or Material 3,5-Diamino-1,2,4-triazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10204999 Certificate TypeCertificate of Analysis Date05/12/2023 Product CodeB22775.06,B22775.22,B22775.14
Lot Number10243302 Certificate TypeCertificate of Analysis Date13/10/2022 Product CodeB22775.06,B22775.22,B22775.14
Lot Number10195678 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB22775.06,B22775.22,B22775.14
Lot NumberUSLF005993 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB22775.06,B22775.22,B22775.14
Lot Number10204999 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB22775.06,B22775.22,B22775.14
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Diamino-1,2,4-triazole, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.