Learn More
3,5-Di(trifluoromethyl)benzoic acid, Tech., Thermo Scientific™
Product Code. 10608765
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10608765 5 g 5g
10587094 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10608765 Supplier Thermo Scientific Maybridge Supplier No. DP01857DE
 Restricted Item
Forventet forsendelse: 16-12-2026
Add to basket
Chemical Identifiers
CAS 725-89-3
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.12
MDL Number MFCD00000388
InChI Key HVFQJWGYVXKLTE-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725
PubChem CID 12889
IUPAC Name 3,5-bis(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Specifications
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CF3)2C6H3CO2H
Quantity 5 g
Formula Weight 258.12
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3, 5-Di(trifluoromethyl)benzoic acid