Få mere at vide
3-(5-Chloropyridine-2-carbamoyl)-2-pyrazinecarboxylic Acid, Mikromol™
Artikelnummer. 30463866
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30463866 4 x 25 mg
30051659 25 mg
30052170 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30463866 Leverandør Mikromol Leverandørnr. MM0149.0500254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-(5-Chloropyridine-2-carbamoyl)-2-pyrazinecarboxylic Acid, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-(5-Chloropyridine-2-carbamoyl)-2-pyrazinecarboxylic Acid
CAS 43200-83-5
Analyt- eller komponentnavne 3-(5-Chloropyridine-2-carbamoyl)-2-pyrazinecarboxylic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C11 H7 Cl N4 O3
Certificeringer/compliance ISO 17025
Synonym 3-(5-Chloropyridine-2-carbamoyl)-2-pyrazinecarboxylic Acid
InChI formel InChI=1S/C11H7ClN4O3/c12-6-1-2-7(15-5-6)16-10(17)8-9(11(18)19)14-4-3-13-8/h1-5H,(H,18,19)(H,15,16,17)
Løsningstype Neat
SMIL OC(=O)c1nccnc1C(=O)Nc2ccc(Cl)cn2
IUPAC navn 3-[(5-chloropyridin-2-yl)carbamoyl]pyrazine-2-carboxylic acid
Molekylvægt (g/mol) 278.65
Mængde 4 x 25 mg
Formel vægt 278.0207 g/mol