Learn More
3,5-Bis(trifluoromethyl)phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Catalog Number p-7024935
Click to view available options
Quantity:
1 g
5 g
25 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
23165-29-9
C9H3F6NS
271.18
MFCD00040838
FXOSSGVJGGNASE-UHFFFAOYSA-N
3,5-bis trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl phenylisothiocyanate, isothiocyanic acid 3,5-bis trifluoromethyl phenyl ester, 3,5-di trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethylphenyl isothiocyanate, benzene, 1-isothiocyanato-3,5-bis trifluoromethyl, pubchem5019, acmc-209g1w
2733395
1-isothiocyanato-3,5-bis(trifluoromethyl)benzene
FC(F)(F)C1=CC(=CC(=C1)N=C=S)C(F)(F)F
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
23165-29-9
C9H3F6NS
271.18
MFCD00040838
FXOSSGVJGGNASE-UHFFFAOYSA-N
3,5-bis trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl phenylisothiocyanate, isothiocyanic acid 3,5-bis trifluoromethyl phenyl ester, 3,5-di trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethylphenyl isothiocyanate, benzene, 1-isothiocyanato-3,5-bis trifluoromethyl, pubchem5019, acmc-209g1w
2733395
1-isothiocyanato-3,5-bis(trifluoromethyl)benzene
FC(F)(F)C1=CC(=CC(=C1)N=C=S)C(F)(F)F
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an active pharmaceutical intermediate.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from alcohols, amines and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from water/moisture.
TRUSTED_SUSTAINABILITY
Density 1.485
Boiling Point 109°C to 110°C (35 mmHg)
Flash Point 108°C (226°F)
Odor Irritating
Refractive Index 1.5
Quantity 1 g
UN Number UN2922
Beilstein 2990816
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 271.19
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenyl isothiocyanate