missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
3,5-Bis(trifluoromethyl)-1H-1,2,4-triazole, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3,5-Bis(trifluoromethyl)-1H-1,2,4-triazole |
| CAS | 709-62-6 |
| Molekylær formel | C4 H F6 N3 |
| Mængde | 25 mg |
| Synonym | 3,5-Bis(trifluoromethyl)-1,2,4-triazole, 3,5-Bis(trifluoromethyl)-s-triazole |
| InChI formel | InChI=1S/C4HF6N3/c5-3(6,7)1-11-2(13-12-1)4(8,9)10/h(H,11,12,13) |
| SMIL | FC(F)(F)c1nnc([nH]1)C(F)(F)F |
| IUPAC navn | 3,5-bis(trifluoromethyl)-4H-1,2,4-triazole |
| Molekylvægt (g/mol) | 205.061 |
| Formel vægt | 205.007 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?