Få mere at vide
3',5'-Bis-O-(tert-butyldimethylsilyl)thymidine-d3, TRC
Artikelnummer. 30339283
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30339283 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339283 Leverandør Toronto Research Chemicals Leverandørnr. TRCB58538250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3',5'-Bis-O-(tert-butyldimethylsilyl)thymidine-d3
CAS 1280736-10-8
Molekylær formel C22H39D3N2O5Si2
Mængde 50 mg
Synonym 3',5'-Bis-O-[(1,1-dimethylethyl)dimethylsilyl]thymidine-d3
InChI formel InChI=1S/C22H42N2O5Si2/c1-15-13-24(20(26)23-19(15)25)18-12-16(29-31(10,11)22(5,6)7)17(28-18)14-27-30(8,9)21(2,3)4/h13,16-18H,12,14H2,1-11H3,(H,23,25,26)/t16?,17-,18+/m0/s1/i1D3
SMIL [2H]C([2H])([2H])C1=CN([C@H]2CC(O[Si](C)(C)C(C)(C)C)[C@H](CO[Si](C)(C)C(C)(C)C)O2)C(=O)NC1=O
IUPAC navn 1-[(2R,5S)-4-[tert-butyl(dimethyl)silyl]oxy-5-[[tert-butyl(dimethyl)silyl]oxymethyl]oxolan-2-yl]-5-(trideuteriomethyl)pyrimidine-2,4-dione
Molekylvægt (g/mol) 473.77
Formel vægt 473.2821