Få mere at vide
3,5-Bis(1,1-dimethylethyl)-4-hydroxy-benzenepropanoic Acid C7-9-Branched Alkyl Esters, TRC
Artikelnummer. 30137622
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137622 2 mg
30135519 1 mg
30009978 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137622 Leverandør Toronto Research Chemicals Leverandørnr. TRCO2774982MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 125643-61-0
Molekylær formel C25H42O3
Molekylvægt (g/mol) 390.61
IUPAC navn octyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate
SMIL CCCCCCCCOC(=O)CCc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C
Tekniske data
Kemisk navn eller materiale 3,5-Bis(1,1-dimethylethyl)-4-hydroxy-benzenepropanoic Acid C7-9-Branched Alkyl Esters
Mængde 2 mg
InChI formel InChI=1S/C25H42O3/c1-8-9-10-11-12-13-16-28-22(26)15-14-19-17-20(24(2,3)4)23(27)21(18-19)25(5,6)7/h17-18,27H,8-16H2,1-7H3
Formel vægt 390.3134