missing translation for 'onlineSavingsMsg'
Få mere at vide

3-[[[5-(Aminocarbonyl)-1-methyl-3-propyl-1H-pyrazol-4-yl]amino]carbonyl]-4-propoxybenzene, TRC

Artikelnummer. 30317842
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317842 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30317842 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6031051G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 3-(((5-(Aminocarbonyl)-1-methyl-3-propyl-1H-pyrazol-4-yl)amino)carbonyl)-4-propoxybenzene
CAS 139756-04-0
Molekylær formel C18H24N4O3
Mængde 1 g
Synonym 1H-Pyrazole-5-carboxamide, 1-methyl-4-[(2-propoxybenzoyl)amino]-3-propyl-, 1-Methyl-4-[(2-propoxybenzoyl)amino]-3-propyl-1H-pyrazole-5-carboxamide, 1-Methyl-4-(2-propoxybenzamido)-3-propylpyrazole-5-carboxamide, 2-Methyl-4-(2-propoxybenzoylamino)-5-propyl-2H-pyrazole-3-carboxamide, 2-Methyl-4-[(2-propoxybenzoyl)amino]-5-propylpyrazole-3-carboxamide, 4-(2-Propoxybenzoylamino)-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
InChI formel InChI=1S/C18H24N4O3/c1-4-8-13-15(16(17(19)23)22(3)21-13)20-18(24)12-9-6-7-10-14(12)25-11-5-2/h6-7,9-10H,4-5,8,11H2,1-3H3,(H2,19,23)(H,20,24)
SMIL CCCOc1ccccc1C(=O)Nc2c(CCC)nn(C)c2C(=O)N
IUPAC navn 2-methyl-4-[(2-propoxybenzoyl)amino]-5-propylpyrazole-3-carboxamide
Molekylvægt (g/mol) 344.41
Formel vægt 344.1848
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.