missing translation for 'onlineSavingsMsg'
Få mere at vide

3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one, 98%, BLD Pharm™

Artikelnummer. 30999313
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
10 g
25 g
100 g
Pakningsstørrelse:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
30999313 100 g 100g
30995977 5 g 5g
30990507 25 g 25g
30990237 1 g 1g
30993816 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30999313 Leverandør BLD Pharm Leverandørnr. BD140581100g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one
CAS 529-44-2
Molekylær formel C15H10O8
MDL nummer MFCD00006827
Mængde 100 g
SMIL OC1=CC(O)=C2C(=O)C(O)=C(OC2=C1)C1=CC(O)=C(O)C(O)=C1
Molekylvægt (g/mol) 318.24
Procent renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.