Få mere at vide
3-{5,7-Dimethylpyrazolo[1,5-a]pyrimidin-6-yl}propanoic Acid, TRC
Artikelnummer. 30376904
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376904 50 mg
30371982 10 mg
30369437 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30376904 Leverandør Toronto Research Chemicals Leverandørnr. TRCD50297850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 832135-73-6
Molekylær formel C11H13N3O2
Molekylvægt (g/mol) 219.24
IUPAC navn 3-(5,7-dimethylpyrazolo[1,5-a]pyrimidin-6-yl)propanoic acid
SMIL Cc1nc2ccnn2c(C)c1CCC(=O)O
Tekniske data
Kemisk navn eller materiale 3-(5,7-Dimethylpyrazolo[1,5-a]pyrimidin-6-yl)propanoic Acid
Mængde 50 mg
InChI formel InChI=1S/C11H13N3O2/c1-7-9(3-4-11(15)16)8(2)14-10(13-7)5-6-12-14/h5-6H,3-4H2,1-2H3,(H,15,16)
Formel vægt 219.1008